Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGTCACTTGGCATCTGGCCATC[A/G]GGCTGGATGCCGTGTTCCAGGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBA1A - tubulin alpha 1a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270399.1 | 529 | Silent Mutation | CCC,CCT | P,P 32 | NP_001257328.1 | |
NM_001270400.1 | 529 | UTR 5 | NP_001257329.1 | |||
NM_006009.3 | 529 | Silent Mutation | CCC,CCT | P,P 32 | NP_006000.2 |