Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGTGGTCCCCAGGAAGCCCCC[G/T]GTGCGGCTGGGAGTCGAACCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603328 | ||||||||||||||||||||
Literature Links: |
MSI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSI1 - musashi RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002442.3 | 1672 | Silent Mutation | ACA,ACC | T,T 304 | NP_002433.1 | |
XM_006719403.3 | 1672 | Silent Mutation | ACA,ACC | T,T 293 | XP_006719466.1 | |
XM_006719404.3 | 1672 | Silent Mutation | ACA,ACC | T,T 166 | XP_006719467.1 | |
XM_011538361.2 | 1672 | Silent Mutation | ACA,ACC | T,T 304 | XP_011536663.1 | |
XM_011538362.2 | 1672 | Silent Mutation | ACA,ACC | T,T 285 | XP_011536664.1 | |
XM_011538363.2 | 1672 | Silent Mutation | ACA,ACC | T,T 274 | XP_011536665.1 | |
XM_011538364.2 | 1672 | Silent Mutation | ACA,ACC | T,T 204 | XP_011536666.1 | |
XM_011538365.2 | 1672 | Silent Mutation | ACA,ACC | T,T 201 | XP_011536667.1 | |
XM_011538366.2 | 1672 | Silent Mutation | ACA,ACC | T,T 201 | XP_011536668.1 | |
XM_011538368.2 | 1672 | Silent Mutation | ACA,ACC | T,T 166 | XP_011536670.1 | |
XM_011538370.2 | 1672 | Silent Mutation | ACA,ACC | T,T 166 | XP_011536672.1 | |
XM_011538371.2 | 1672 | Silent Mutation | ACA,ACC | T,T 166 | XP_011536673.1 |