Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTTCACTGTGACACCATGCAGA[A/G]TCGGAACTCTAACTGGTATTAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F7 - E2F transcription factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203394.2 | 3172 | UTR 3 | NP_976328.2 | |||
XM_011537966.2 | 3172 | UTR 3 | XP_011536268.1 | |||
XM_011537969.2 | 3172 | UTR 3 | XP_011536271.1 |