Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTTCGAGACAGCCTGAGCAATA[A/G]TGGATCCTGTATCTGGGGAAAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGES3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGES3 - prostaglandin E synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282601.1 | Intron | NP_001269530.1 | ||||
NM_001282602.1 | Intron | NP_001269531.1 | ||||
NM_001282603.1 | Intron | NP_001269532.1 | ||||
NM_001282604.1 | Intron | NP_001269533.1 | ||||
NM_001282605.1 | Intron | NP_001269534.1 | ||||
NM_006601.6 | Intron | NP_006592.3 | ||||
XM_005268576.4 | Intron | XP_005268633.1 | ||||
XM_006719199.2 | Intron | XP_006719262.1 | ||||
XM_011537773.2 | Intron | XP_011536075.1 | ||||
XM_011537774.2 | Intron | XP_011536076.1 | ||||
XM_017018716.1 | Intron | XP_016874205.1 |