Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGAATCTCTCTTGGGTCTTAC[C/G]GTCTACGGGAGTAATGATCAAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PWP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PWP1 - PWP1 homolog, endonuclein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317962.1 | 381 | Silent Mutation | ACC,ACG | T,T 57 | NP_001304891.1 | |
NM_001317963.1 | 381 | UTR 5 | NP_001304892.1 | |||
NM_007062.2 | 381 | Silent Mutation | ACC,ACG | T,T 119 | NP_008993.1 |