Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAAACTCACGCATATGCACAAA[C/T]GCACATCAACAAACACACAAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615335 MIM: 614478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS5 - ceramide synthase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281731.1 | 1652 | UTR 3 | NP_001268660.1 | |||
NM_147190.3 | 1652 | UTR 3 | NP_671723.1 | |||
XM_005269220.2 | 1652 | UTR 3 | XP_005269277.1 | |||
XM_006719687.3 | 1652 | UTR 3 | XP_006719750.1 | |||
XM_011538975.2 | 1652 | UTR 3 | XP_011537277.1 | |||
XM_017020202.1 | 1652 | UTR 3 | XP_016875691.1 | |||
XM_017020203.1 | 1652 | UTR 3 | XP_016875692.1 | |||
XM_017020204.1 | 1652 | Intron | XP_016875693.1 | |||
XM_017020205.1 | 1652 | UTR 3 | XP_016875694.1 |
COX14 - COX14, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |