Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCTCATGGACAGAAACTTCA[C/T]TTGGCGTGAGGTGGGAAGAGCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601478 MIM: 616496 | ||||||||||||||||||||
Literature Links: |
MYO1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO1A - myosin IA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130963.1 | 1242 | Missense Mutation | AAT,AGT | N,S 399 | NP_001124435.1 | |
NM_015257.2 | 1242 | Missense Mutation | AAT,AGT | N,S 326 | NP_056072.2 | |
XM_011538056.2 | 1242 | Missense Mutation | AAT,AGT | N,S 447 | XP_011536358.1 | |
XM_011538057.2 | 1242 | Missense Mutation | AAT,AGT | N,S 407 | XP_011536359.1 | |
XM_011538058.2 | 1242 | Missense Mutation | AAT,AGT | N,S 374 | XP_011536360.1 | |
XM_011538059.1 | 1242 | Missense Mutation | AAT,AGT | N,S 228 | XP_011536361.1 | |
XM_011538060.2 | 1242 | Intron | XP_011536362.1 | |||
XM_017019077.1 | 1242 | Missense Mutation | AAT,AGT | N,S 407 | XP_016874566.1 | |
XM_017019078.1 | 1242 | Missense Mutation | AAT,AGT | N,S 228 | XP_016874567.1 | |
XM_017019079.1 | 1242 | Missense Mutation | AAT,AGT | N,S 208 | XP_016874568.1 |