Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGATGACTGGGACTCCTGTCCT[C/T]GTGGCCCTCGGGGACTTCCTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL42 - kelch like family member 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020782.1 | 962 | Silent Mutation | CTC,CTT | L,L 196 | NP_065833.1 | |
XM_017019698.1 | 962 | Silent Mutation | CTC,CTT | L,L 196 | XP_016875187.1 |
MANSC4 - MANSC domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |