Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGAATTATACTACTGTGGTTG[A/C]GTTAGGTACTTTATATTAGATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGAM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGAM5 - PGAM family member 5, mitochondrial serine/threonine protein phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170543.1 | Intron | NP_001164014.1 | ||||
NM_001170544.1 | Intron | NP_001164015.1 | ||||
NM_138575.3 | Intron | NP_612642.2 |
PXMP2 - peroxisomal membrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |