Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTATTATCCTTTTCCTTGTCTT[C/T]CATCTTCTTTGCCTCTCCAGGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605936 | ||||||||||||||||||||
Literature Links: |
BIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BIN2 - bridging integrator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290007.1 | Intron | NP_001276936.1 | ||||
NM_001290008.1 | Intron | NP_001276937.1 | ||||
NM_001290009.1 | Intron | NP_001276938.1 | ||||
NM_016293.3 | Intron | NP_057377.3 | ||||
XM_005268957.3 | Intron | XP_005269014.2 | ||||
XM_005268958.3 | Intron | XP_005269015.1 | ||||
XM_017019422.1 | Intron | XP_016874911.1 |