Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGATCCTGAGATGGTGTACACA[A/G]TTCTCCAACATCGAGACTACCACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615123 | ||||||||||||||||||||
Literature Links: |
ANKRD13A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD13A - ankyrin repeat domain 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033121.1 | 536 | Missense Mutation | ATT,GTT | I,V 93 | NP_149112.1 | |
XM_005253980.2 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_005254037.1 | |
XM_005253981.3 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_005254038.1 | |
XM_005253982.3 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_005254039.1 | |
XM_005253984.1 | 536 | Missense Mutation | ATT,GTT | I,V 5 | XP_005254041.1 | |
XM_011538937.1 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_011537239.1 | |
XM_011538938.2 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_011537240.1 | |
XM_017020157.1 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_016875646.1 | |
XM_017020158.1 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_016875647.1 | |
XM_017020159.1 | 536 | Missense Mutation | ATT,GTT | I,V 5 | XP_016875648.1 | |
XM_017020160.1 | 536 | Missense Mutation | ATT,GTT | I,V 5 | XP_016875649.1 | |
XM_017020161.1 | 536 | Missense Mutation | ATT,GTT | I,V 5 | XP_016875650.1 | |
XM_017020162.1 | 536 | Missense Mutation | ATT,GTT | I,V 93 | XP_016875651.1 | |
XM_017020163.1 | 536 | UTR 5 | XP_016875652.1 |