Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAACCCCTAGAGGGCAAGTACGA[C/G]TGGCAAGAGGTGGAGAAGGGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612456 MIM: 600778 MIM: 602927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOLD1 - apolipoprotein L domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKN1B - cyclin dependent kinase inhibitor 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004064.4 | 795 | Missense Mutation | GAC,GAG | D,E 75 | NP_004055.1 |
GPR19 - G protein-coupled receptor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006143.2 | 795 | Intron | NP_006134.1 |