Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACAACAAAAAACCCAAATGGAA[C/T]CTATAAAGGGAAGACAGATATATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL14 - F-box and leucine rich repeat protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152441.2 | 2106 | UTR 3 | NP_689654.1 | |||
XM_017018875.1 | 2106 | Intron | XP_016874364.1 | |||
XM_017018876.1 | 2106 | Intron | XP_016874365.1 |