Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTACCAGTCACACCTCATGACA[A/G]TTGCCTCTTTGTTTATTATGCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPC3 - actin related protein 2/3 complex subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM216A - family with sequence similarity 216 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPN3 - GPN-loop GTPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164372.1 | Intron | NP_001157844.1 | ||||
NM_001164373.1 | Intron | NP_001157845.1 | ||||
NM_016301.3 | Intron | NP_057385.3 | ||||
XM_005253896.4 | Intron | XP_005253953.3 | ||||
XM_017019394.1 | Intron | XP_016874883.1 |