Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCTTTCTCTACTCCTAGGAGTT[C/G]CTGTCCAACCCTCCAGCTGGTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESPL1 - extra spindle pole bodies like 1, separase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012291.4 | 178 | Missense Mutation | TTC,TTG | F,L 29 | NP_036423.4 | |
XM_006719705.3 | 178 | Missense Mutation | TTC,TTG | F,L 29 | XP_006719768.1 | |
XM_011539024.2 | 178 | Missense Mutation | TTC,TTG | F,L 29 | XP_011537326.1 | |
XM_011539025.2 | 178 | Missense Mutation | TTC,TTG | F,L 29 | XP_011537327.1 | |
XM_017020253.1 | 178 | UTR 5 | XP_016875742.1 |