Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTTTCCATCGGAAGATATTCT[C/T]TTGGTAACAGCTTCTCCTGTTATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PYROXD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PYROXD1 - pyridine nucleotide-disulphide oxidoreductase domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024854.3 | 244 | Silent Mutation | NP_079130.2 | |||
XM_006719153.3 | 244 | Silent Mutation | CTC,CTT | L,L 39 | XP_006719216.1 | |
XM_017019976.1 | 244 | Intron | XP_016875465.1 |