Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACTGTCCATCTCACCTCTGTC[A/G]GATGCCAGCCTGGACCTGCAGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610576 MIM: 165220 MIM: 612031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP9 - Rho GTPase activating protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLI1 - GLI family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160045.1 | 591 | UTR 5 | NP_001153517.1 | |||
NM_001167609.1 | 591 | Silent Mutation | TCA,TCG | S,S 48 | NP_001161081.1 | |
NM_005269.2 | 591 | Silent Mutation | TCA,TCG | S,S 89 | NP_005260.1 | |
XM_011538189.2 | 591 | Silent Mutation | TCA,TCG | S,S 89 | XP_011536491.1 | |
XM_011538190.1 | 591 | Silent Mutation | TCA,TCG | S,S 89 | XP_011536492.1 |
INHBE - inhibin beta E subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |