Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCTCCCGCAAGGACCTGATCGG[C/T]CACGGCGCCTTCGCGGTGGTCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603168 | ||||||||||||||||||||
Literature Links: |
ULK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ULK1 - unc-51 like autophagy activating kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003565.2 | 337 | Silent Mutation | GGC,GGT | G,G 23 | NP_003556.1 | |
XM_011538798.2 | 337 | Silent Mutation | GGC,GGT | G,G 23 | XP_011537100.1 | |
XM_011538799.2 | 337 | Silent Mutation | GGC,GGT | G,G 23 | XP_011537101.1 |