Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCGAAGTCTGCAGCAAGGATAG[C/T]TTCAGCATCATCATCCTATTTTTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164060 | ||||||||||||||||||||
Literature Links: |
NAP1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAP1L1 - nucleosome assembly protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719425.3 | 1351 | Missense Mutation | ACT,GCT | T,A 255 | XP_006719488.1 | |
XM_011538390.2 | 1351 | Missense Mutation | ACT,GCT | T,A 318 | XP_011536692.2 | |
XM_011538393.2 | 1351 | Missense Mutation | ACT,GCT | T,A 318 | XP_011536695.1 | |
XM_017019338.1 | 1351 | Missense Mutation | ACT,GCT | T,A 318 | XP_016874827.1 | |
XM_017019339.1 | 1351 | Missense Mutation | ACT,GCT | T,A 255 | XP_016874828.1 | |
XM_017019340.1 | 1351 | Missense Mutation | ACT,GCT | T,A 255 | XP_016874829.1 |