Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGTAGAGAGAAAAGCATCAGAG[G/T]TATCCAGTGGGCTGGTTTCGTGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605246 | ||||||||||||||||||||
Literature Links: |
C3AR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C3AR1 - complement component 3a receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326475.1 | 1058 | Missense Mutation | AAC,ACC | N,T 285 | NP_001313404.1 | |
NM_001326477.1 | 1058 | Missense Mutation | AAC,ACC | N,T 285 | NP_001313406.1 | |
NM_004054.3 | 1058 | Missense Mutation | AAC,ACC | N,T 285 | NP_004045.1 |
FOXJ2 - forkhead box J2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |