Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGTTTAGCAAAGATGGCTAAAG[C/T]ATGGGCAAACCAGTGCAAATTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607724 MIM: 610395 | ||||||||||||||||||||
Literature Links: |
CAPS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLIPR1L1 - GLI pathogenesis related 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304964.1 | 3094 | Missense Mutation | GCA,GTA | A,V 70 | NP_001291893.1 | |
NM_152779.3 | 3094 | Missense Mutation | GCA,GTA | A,V 70 | NP_689992.1 | |
XM_011538134.2 | 3094 | Missense Mutation | GCA,GTA | A,V 155 | XP_011536436.2 | |
XM_017019134.1 | 3094 | Intron | XP_016874623.1 |