Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTCTGTAGTACTAAAAGAAAG[A/G]TAATATTTTCTCATTATCTTCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRIQ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRIQ1 - leucine rich repeats and IQ motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079910.1 | Intron | NP_001073379.1 | ||||
XM_011538817.2 | Intron | XP_011537119.1 | ||||
XM_011538818.2 | Intron | XP_011537120.1 | ||||
XM_011538819.2 | Intron | XP_011537121.1 | ||||
XM_011538823.2 | Intron | XP_011537125.1 | ||||
XM_011538824.2 | Intron | XP_011537126.1 | ||||
XM_011538825.2 | Intron | XP_011537127.1 | ||||
XM_017020019.1 | Intron | XP_016875508.1 |
TSPAN19 - tetraspanin 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |