Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTAGCTGTAGACAGAAACAAA[A/C]ATACGATTATTATTTTACTTAGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609229 MIM: 603679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT4 - nudix hydrolase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2N - ubiquitin conjugating enzyme E2 N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003348.3 | Intron | NP_003339.1 | ||||
XM_011538719.2 | Intron | XP_011537021.1 | ||||
XM_017019918.1 | Intron | XP_016875407.1 |