Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATATTTCTATATATAATATGATC[A/G]GGCCAATATTGATAGTATAGTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATS2 - spermatogenesis associated serine rich 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293285.1 | Intron | NP_001280214.1 | ||||
NM_001293286.1 | Intron | NP_001280215.1 | ||||
NM_023071.3 | Intron | NP_075559.2 | ||||
XM_011538690.2 | Intron | XP_011536992.1 | ||||
XM_011538691.2 | Intron | XP_011536993.1 | ||||
XM_017019840.1 | Intron | XP_016875329.1 |