Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTGGATTAAAATCATGGTCAAC[C/T]AGATGTTGCTTTTGCCTGGGAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602287 MIM: 612755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3657 - microRNA 3657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA25 - N(alpha)-acetyltransferase 25, NatB auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024953.3 | 3712 | UTR 3 | NP_079229.2 | |||
XM_005253938.1 | 3712 | UTR 3 | XP_005253995.1 | |||
XM_006719606.2 | 3712 | UTR 3 | XP_006719669.1 | |||
XM_017019977.1 | 3712 | UTR 3 | XP_016875466.1 |