Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAATATTTTTACCAAAATTTGGA[G/T]ATGGCATCTCGCTCGGTATTTTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607971 | ||||||||||||||||||||
Literature Links: |
SLC6A15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC6A15 - solute carrier family 6 member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146335.2 | 2555 | Missense Mutation | TAT,TCT | Y,S 580 | NP_001139807.1 | |
NM_018057.6 | 2555 | Intron | NP_060527.2 | |||
NM_182767.5 | 2555 | Missense Mutation | TAT,TCT | Y,S 687 | NP_877499.1 | |
XM_011538525.2 | 2555 | Intron | XP_011536827.1 |