Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGATGGCCTGGTTATTGACCAGC[A/T]TGAGATATTTGTGGACTACGAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RILPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RILPL1 - Rab interacting lysosomal protein like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP35 - small nuclear ribonucleoprotein U11/U12 subunit 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022717.3 | 1658 | Missense Mutation | CAT,CTT | H,L 120 | NP_073208.1 | |
NM_180699.3 | 1658 | Missense Mutation | CAT,CTT | H,L 125 | NP_851030.1 | |
XM_006719204.3 | 1658 | Missense Mutation | CAT,CTT | H,L 120 | XP_006719267.1 | |
XM_011537780.2 | 1658 | Missense Mutation | CAT,CTT | H,L 181 | XP_011536082.1 | |
XM_017018724.1 | 1658 | Missense Mutation | CAT,CTT | H,L 125 | XP_016874213.1 |