Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCAGCCGCCTTACTCTGAGA[C/T]CTCCGACTCCTGCGAGAAGTACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611161 | ||||||||||||||||||||
Literature Links: |
KRT80 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT80 - keratin 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081492.1 | 1481 | UTR 3 | NP_001074961.1 | |||
NM_182507.2 | 1481 | Missense Mutation | ATC,GTC | I,V 450 | NP_872313.2 | |
XM_005268676.3 | 1481 | Missense Mutation | ATC,GTC | I,V 485 | XP_005268733.1 |