Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTTGGCTTTTTCTCTGGACAC[C/T]TTGAAGCTCAGAGGATAGACGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR19 - G protein-coupled receptor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006143.2 | 3220 | Silent Mutation | AAA,AAG | K,K 174 | NP_006134.1 | |
XM_011520623.2 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_011518925.1 | |
XM_011520624.2 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_011518926.1 | |
XM_011520625.2 | 3220 | Silent Mutation | AAA,AAG | K,K 174 | XP_011518927.1 | |
XM_017019212.1 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_016874701.1 | |
XM_017019213.1 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_016874702.1 | |
XM_017019214.1 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_016874703.1 | |
XM_017019215.1 | 3220 | Silent Mutation | AAA,AAG | K,K 193 | XP_016874704.1 | |
XM_017019216.1 | 3220 | Silent Mutation | AAA,AAG | K,K 174 | XP_016874705.1 |