Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTGGGTTGAGGGCAGCAGTGG[A/G]TGGAAGGACACCAAAGGTAAACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605423 MIM: 610007 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHH - desert hedgehog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021044.3 | 3387 | UTR 3 | NP_066382.1 | |||
XM_017019380.1 | 3387 | UTR 3 | XP_016874869.1 | |||
XM_017019381.1 | 3387 | UTR 3 | XP_016874870.1 |
LMBR1L - limb development membrane protein 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |