Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTGCAGGATGTCAGACAGTGA[C/T]ACAATTCCCTTGACCACATCATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610588 MIM: 602742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDN - dendrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAG1 - protein kinase AMP-activated non-catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206709.1 | 1045 | Silent Mutation | GTA,GTG | V,V 322 | NP_001193638.1 | |
NM_001206710.1 | 1045 | Silent Mutation | GTA,GTG | V,V 281 | NP_001193639.1 | |
NM_002733.4 | 1045 | Silent Mutation | GTA,GTG | V,V 313 | NP_002724.1 | |
XM_005269019.4 | 1045 | Silent Mutation | GTA,GTG | V,V 350 | XP_005269076.1 | |
XM_005269020.2 | 1045 | Silent Mutation | GTA,GTG | V,V 229 | XP_005269077.1 | |
XM_006719499.2 | 1045 | Silent Mutation | GTA,GTG | V,V 281 | XP_006719562.1 | |
XM_011538562.2 | 1045 | Silent Mutation | GTA,GTG | V,V 281 | XP_011536864.1 | |
XM_017019622.1 | 1045 | Silent Mutation | GTA,GTG | V,V 229 | XP_016875111.1 | |
XM_017019623.1 | 1045 | Silent Mutation | GTA,GTG | V,V 229 | XP_016875112.1 |