Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTCCCAAACCTGAAAAACCCA[C/T]GTCAGTTTGTCTGATATGTATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DENND5B - DENN domain containing 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308339.1 | 9421 | UTR 3 | NP_001295268.1 | |||
NM_144973.3 | 9421 | UTR 3 | NP_659410.3 | |||
XM_005253326.2 | 9421 | UTR 3 | XP_005253383.1 | |||
XM_005253327.3 | 9421 | Intron | XP_005253384.1 | |||
XM_005253330.3 | 9421 | Intron | XP_005253387.1 | |||
XM_006719040.3 | 9421 | Intron | XP_006719103.1 | |||
XM_011520590.2 | 9421 | Intron | XP_011518892.1 | |||
XM_017018896.1 | 9421 | UTR 3 | XP_016874385.1 | |||
XM_017018897.1 | 9421 | Intron | XP_016874386.1 | |||
XM_017018898.1 | 9421 | Intron | XP_016874387.1 | |||
XM_017018899.1 | 9421 | Intron | XP_016874388.1 |