Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGAGAGTGTGTAGGAGTCCTG[A/G]TAATCTTGAAGCAGTTTGACCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606365 | ||||||||||||||||||||
Literature Links: |
GLS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLS2 - glutaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280796.1 | 1891 | Silent Mutation | TAC,TAT | Y,Y 299 | NP_001267725.1 | |
NM_001280797.1 | 1891 | Silent Mutation | TAC,TAT | Y,Y 310 | NP_001267726.1 | |
NM_001280798.1 | 1891 | Silent Mutation | TAC,TAT | Y,Y 310 | NP_001267727.1 | |
NM_013267.3 | 1891 | Silent Mutation | TAC,TAT | Y,Y 575 | NP_037399.2 | |
XM_005268797.1 | 1891 | Silent Mutation | TAC,TAT | Y,Y 539 | XP_005268854.1 | |
XM_017019180.1 | 1891 | Silent Mutation | TAC,TAT | Y,Y 310 | XP_016874669.1 |
SPRYD4 - SPRY domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207344.3 | 1891 | Intron | NP_997227.1 |