Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGTACCTTTAGCTTTTTTTCCA[C/T]TTGACTTCTCATCAGGCTGGTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602601 | ||||||||||||||||||||
Literature Links: |
OLR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLR1 - oxidized low density lipoprotein receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172632.1 | 173 | Missense Mutation | AAT,AGT | N,S 20 | NP_001166103.1 | |
NM_001172633.1 | 173 | Missense Mutation | AAT,AGT | N,S 20 | NP_001166104.1 | |
NM_002543.3 | 173 | Missense Mutation | AAT,AGT | N,S 20 | NP_002534.1 |
TMEM52B - transmembrane protein 52B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079815.1 | 173 | Intron | NP_001073283.1 | |||
NM_153022.3 | 173 | Intron | NP_694567.1 | |||
XM_006719029.2 | 173 | Intron | XP_006719092.1 |