Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTAACACATTAACATTATACCAT[A/G]CACTGATTTTATTTTCACAACCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TSPAN19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPAN19 - tetraspanin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100917.1 | 656 | Missense Mutation | GCA,GTA | A,V 206 | NP_001094387.1 | |
XM_005268669.4 | 656 | Missense Mutation | GCA,GTA | A,V 206 | XP_005268726.1 | |
XM_017018862.1 | 656 | Missense Mutation | GCA,GTA | A,V 206 | XP_016874351.1 | |
XM_017018863.1 | 656 | Missense Mutation | GCA,GTA | A,V 204 | XP_016874352.1 | |
XM_017018864.1 | 656 | UTR 3 | XP_016874353.1 |