Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTCACTATGTGGCTCTACCTGG[C/T]GGCCTTCGTGGGCCTGTACTACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B6 - hydroxysteroid 17-beta dehydrogenase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003725.3 | 357 | Missense Mutation | GCG,GTG | A,V 6 | NP_003716.2 | |
XM_005269207.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_005269264.1 | |
XM_005269208.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_005269265.1 | |
XM_005269209.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_005269266.1 | |
XM_006719672.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_006719735.1 | |
XM_011538925.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_011537227.1 | |
XM_011538926.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_011537228.1 | |
XM_011538927.1 | 357 | Missense Mutation | GCG,GTG | A,V 6 | XP_011537229.1 |