Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACTGTCTTTACAGCAGTTATC[A/G]CACATTTTGTTACATGCTTCTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600537 | ||||||||||||||||||||
Literature Links: |
PYROXD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PYROXD1 - pyridine nucleotide-disulphide oxidoreductase domain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RECQL - RecQ like helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002907.3 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | NP_002898.2 | |
NM_032941.2 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | NP_116559.1 | |
XM_005253461.3 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | XP_005253518.1 | |
XM_005253462.4 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | XP_005253519.1 | |
XM_005253463.3 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | XP_005253520.1 | |
XM_005253464.3 | 1844 | Silent Mutation | TGC,TGT | C,C 475 | XP_005253521.1 |