Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAAATAGTTACCTGGATATATC[G/T]TTTCACTTTTTGAATATGTTTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C12orf50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf50 - chromosome 12 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152589.1 | 1619 | Missense Mutation | AAG,ACG | K,T 403 | NP_689802.1 | |
XM_011537985.1 | 1619 | Intron | XP_011536287.1 | |||
XM_017018886.1 | 1619 | Missense Mutation | AAG,ACG | K,T 402 | XP_016874375.1 | |
XM_017018887.1 | 1619 | Missense Mutation | AAG,ACG | K,T 457 | XP_016874376.1 | |
XM_017018888.1 | 1619 | Missense Mutation | AAG,ACG | K,T 364 | XP_016874377.1 |