Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGTACAGCTCTGTCAGCTGTTGG[C/T]GCATGACCTCCAGGGTCCGGTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164017 MIM: 601490 | ||||||||||||||||||||
Literature Links: |
HNRNPA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPA1 - heterogeneous nuclear ribonucleoprotein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFE2 - nuclear factor, erythroid 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136023.2 | 1391 | Missense Mutation | CAC,CGC | H,R 323 | NP_001129495.1 | |
NM_001261461.1 | 1391 | Missense Mutation | CAC,CGC | H,R 323 | NP_001248390.1 | |
NM_006163.2 | 1391 | Missense Mutation | CAC,CGC | H,R 323 | NP_006154.1 | |
XM_005268906.4 | 1391 | Missense Mutation | CAC,CGC | H,R 323 | XP_005268963.1 |