Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGGGCGGCATCGGCTCTGCTG[A/C]TGCGGAGCCCCCGCCTGCCCGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611911 MIM: 611684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISCU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISCU - iron-sulfur cluster assembly enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301140.1 | 1212 | Silent Mutation | ATG,CTG | M,L 18 | NP_001288069.1 | |
NM_001301141.1 | 1212 | Silent Mutation | ATG,CTG | M,L 18 | NP_001288070.1 | |
NM_001320042.1 | 1212 | Silent Mutation | ATG,CTG | M,L 18 | NP_001306971.1 | |
NM_014301.4 | 1212 | UTR 5 | NP_055116.1 | |||
NM_213595.3 | 1212 | Silent Mutation | ATG,CTG | M,L 18 | NP_998760.1 | |
XM_011538100.2 | 1212 | UTR 5 | XP_011536402.1 | |||
XM_011538101.2 | 1212 | Silent Mutation | ATG,CTG | M,L 18 | XP_011536403.1 |
SART3 - squamous cell carcinoma antigen recognized by T-cells 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |