Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATAAGTTCATCAAACTACTTCC[A/G]TCTGAAAAATCCTGATCTCAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK38L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK38L - serine/threonine kinase 38 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015000.3 | 118 | Intron | NP_055815.1 | |||
XM_005253342.3 | 118 | Intron | XP_005253399.1 | |||
XM_006719058.3 | 118 | Intron | XP_006719121.1 | |||
XM_006719059.3 | 118 | Intron | XP_006719122.1 | |||
XM_011520613.2 | 118 | Intron | XP_011518915.1 | |||
XM_017019036.1 | 118 | UTR 5 | XP_016874525.1 |