Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGAGTGGCTTGCGTGACACTGC[A/G]TTGTTTCTTCAGATCATACTTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
UHRF1BP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UHRF1BP1L - UHRF1 binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006947.1 | 2881 | Intron | NP_001006948.1 | |||
NM_015054.1 | 2881 | Missense Mutation | CGC,TGC | R,C 1393 | NP_055869.1 | |
XM_005268738.2 | 2881 | Missense Mutation | CGC,TGC | R,C 1345 | XP_005268795.1 | |
XM_005268739.3 | 2881 | Missense Mutation | CGC,TGC | R,C 1332 | XP_005268796.1 | |
XM_011538054.2 | 2881 | Missense Mutation | CGC,TGC | R,C 911 | XP_011536356.1 | |
XM_017019047.1 | 2881 | Missense Mutation | CGC,TGC | R,C 1393 | XP_016874536.1 | |
XM_017019048.1 | 2881 | Missense Mutation | CGC,TGC | R,C 1284 | XP_016874537.1 | |
XM_017019049.1 | 2881 | Intron | XP_016874538.1 | |||
XM_017019050.1 | 2881 | Missense Mutation | CGC,TGC | R,C 940 | XP_016874539.1 | |
XM_017019051.1 | 2881 | Missense Mutation | CGC,TGC | R,C 940 | XP_016874540.1 |