Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGGAACCTTGAGATTGTGCTCA[C/T]GGGACACAATGCCGACCTCTCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190151 | ||||||||||||||||||||
Literature Links: |
ERBB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERBB3 - erb-b2 receptor tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005915.1 | 396 | Missense Mutation | ACG,ATG | T,M 68 | NP_001005915.1 | |
NM_001982.3 | 396 | Missense Mutation | ACG,ATG | T,M 68 | NP_001973.2 |