Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTCATCCTCATTTCTGGAGA[C/G]AGTGGGGCAGGGAAAACAGAGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO1H - myosin IH | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101421.3 | 760 | Missense Mutation | GAC,GAG | D,E 106 | NP_001094891.3 | |
XM_011538223.2 | 760 | Missense Mutation | GAC,GAG | D,E 106 | XP_011536525.1 | |
XM_017019207.1 | 760 | Missense Mutation | GAC,GAG | D,E 106 | XP_016874696.1 |