Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCTTGACATGTCTTCCCTTAG[C/T]TATACCCCTGGTTCTCCACATCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605307 MIM: 162330 | ||||||||||||||||||||
Literature Links: |
GPR182 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR182 - G protein-coupled receptor 182 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBCBP - HBcAg-binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAC3 - tachykinin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB39 - zinc finger and BTB domain containing 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014830.2 | 3928 | UTR 3 | NP_055645.1 |