Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACACATTGAGGCAGTATGTCCC[C/T]GAGCCACCCTTCAGTATCTGGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125855 MIM: 155550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKA - diacylglycerol kinase alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMEL - premelanosome protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200053.1 | 1611 | Silent Mutation | TCA,TCG | S,S 476 | NP_001186982.1 | |
NM_001200054.1 | 1611 | Silent Mutation | TCA,TCG | S,S 562 | NP_001186983.1 | |
NM_001320121.1 | 1611 | Silent Mutation | TCA,TCG | S,S 520 | NP_001307050.1 | |
NM_001320122.1 | 1611 | Silent Mutation | TCA,TCG | S,S 520 | NP_001307051.1 | |
NM_006928.4 | 1611 | Silent Mutation | TCA,TCG | S,S 562 | NP_008859.1 | |
XM_006719569.1 | 1611 | Silent Mutation | TCA,TCG | S,S 562 | XP_006719632.1 | |
XM_011538685.1 | 1611 | Silent Mutation | TCA,TCG | S,S 562 | XP_011536987.1 |