Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGGCCTTAATTTTCTGTTCATG[G/T]ACTGTTAAAAGTAAAACCAAACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602692 MIM: 612817 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIPR1 - GLI pathogenesis related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006851.2 | 1468 | UTR 3 | NP_006842.2 | |||
XM_011537775.2 | 1468 | UTR 3 | XP_011536077.1 | |||
XM_017018721.1 | 1468 | UTR 3 | XP_016874210.1 |
KRR1 - KRR1, small subunit processome component homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007043.6 | 1468 | UTR 3 | NP_008974.5 |