Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACGTTTCTGCAGCAAAGTGGGC[C/T]GAGCGCAGTACGCACAGGTGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614274 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SBNO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SBNO1 - strawberry notch homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167856.1 | 8025 | UTR 3 | NP_001161328.1 | |||
NM_018183.3 | 8025 | UTR 3 | NP_060653.3 | |||
XM_005253572.3 | 8025 | UTR 3 | XP_005253629.1 | |||
XM_005253573.4 | 8025 | UTR 3 | XP_005253630.1 | |||
XM_005253575.3 | 8025 | UTR 3 | XP_005253632.1 | |||
XM_005253576.3 | 8025 | UTR 3 | XP_005253633.1 | |||
XM_006719473.3 | 8025 | UTR 3 | XP_006719536.1 | |||
XM_006719474.3 | 8025 | UTR 3 | XP_006719537.1 | |||
XM_011538533.2 | 8025 | UTR 3 | XP_011536835.1 | |||
XM_011538534.2 | 8025 | UTR 3 | XP_011536836.1 | |||
XM_011538535.2 | 8025 | UTR 3 | XP_011536837.1 | |||
XM_017019556.1 | 8025 | UTR 3 | XP_016875045.1 | |||
XM_017019557.1 | 8025 | UTR 3 | XP_016875046.1 | |||
XM_017019558.1 | 8025 | UTR 3 | XP_016875047.1 | |||
XM_017019559.1 | 8025 | UTR 3 | XP_016875048.1 |