Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGATATTAAGTACCCATTTAAAG[C/T]GATCATTTTATGGAAAGATAATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609229 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT4 - nudix hydrolase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301022.1 | Intron | NP_001287951.1 | ||||
NM_001301023.1 | Intron | NP_001287952.1 | ||||
NM_001301024.1 | Intron | NP_001287953.1 | ||||
NM_019094.5 | Intron | NP_061967.3 | ||||
NM_199040.3 | Intron | NP_950241.1 | ||||
XM_011537794.2 | Intron | XP_011536096.1 | ||||
XM_011537795.2 | Intron | XP_011536097.1 | ||||
XM_011537796.2 | Intron | XP_011536098.1 |