Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGACACTCCTTCACATGTCTGC[A/G]CGAACAAAAGAAGCAAATACTCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601439 | ||||||||||||||||||||
Literature Links: |
ABCC9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCC9 - ATP binding cassette subfamily C member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005691.3 | 5444 | Intron | NP_005682.2 | |||
NM_020297.3 | 5444 | Silent Mutation | NP_064693.2 | |||
XM_005253284.3 | 5444 | Silent Mutation | XP_005253341.1 | |||
XM_005253286.3 | 5444 | Silent Mutation | XP_005253343.1 | |||
XM_005253287.4 | 5444 | UTR 3 | XP_005253344.1 | |||
XM_005253288.3 | 5444 | Silent Mutation | XP_005253345.1 | |||
XM_005253289.3 | 5444 | Silent Mutation | XP_005253346.1 | |||
XM_005253290.3 | 5444 | Silent Mutation | XP_005253347.1 | |||
XM_006719025.3 | 5444 | UTR 3 | XP_006719088.1 | |||
XM_011520545.2 | 5444 | Silent Mutation | XP_011518847.1 |